Assay Details
Target Gene Details
Entrez Gene ID: | 22223 |
Gene Name: | ubiquitin carboxy-terminal hydrolase L1 |
Gene Aliases: |
AW822034, C88048, PGP 9.5, PGP9.5, R75593, UCH-L1, UCHL-1, gad |
Location: |
Chr.5:66676121-66687234 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uchl1 | NM_011670.2 | 4 | 456 | NP_035800.2 |
| AB025313.1 | 4 | 307 | BAA84083.1 | |
| AF172334.1 | 4 | 295 | AAD51029.1 | |
| AK004066.1 | 4 | 333 | ||
| AK013729.1 | 4 | 336 | BAB28976.1 | |
| AK170728.1 | 2 | 294 | BAE41984.1 | |
| BC039177.1 | 4 | 322 | AAH39177.1 | |
| CX241178.1 | 4 | 456 | ||
| JX050004.1 | 2 | 260 |
More Information
Set Membership: |
|

Genomic Map