Assay Details
Target Gene Details
Entrez Gene ID: | 75788 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 1 |
Gene Aliases: |
4930431E10Rik, mKIAA1625 |
Location: |
Chr.5:144876495-144965830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smurf1 | NM_001038627.1 | NP_001033716.1 | ||
| NM_029438.3 | NP_083714.3 | |||
| AK015264.1 | BAB29770.2 | |||
| AK034736.1 | BAC28813.1 | |||
| AK122534.1 | 5 | 644 | BAC65816.1 | |
| AK154491.1 | BAE32623.1 | |||
| BC029097.1 | AAH29097.1 |
More Information
Set Membership: |
|

Genomic Map