Assay Details
Target Gene Details
Entrez Gene ID: | 94215 |
Gene Name: | UDP glucuronosyltransferase 2 family, polypeptide A1 |
Gene Aliases: |
- |
Location: |
Chr.5:87459490-87490871 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt2a1 | NM_053184.2 | NP_444414.2 | ||
| AF184901.1 | AAG17003.1 | |||
| AK140757.1 | BAE24468.1 | |||
| BC048926.1 | AAH48926.1 |
Target Gene Details
Entrez Gene ID: | 552899 |
Gene Name: | UDP glucuronosyltransferase 2 family, polypeptide A2 |
Gene Aliases: |
- |
Location: |
Chr.5:87459493-87482258 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt2a2 | NM_001024148.1 | NP_001019319.1 | ||
| BC058786.1 | AAH58786.1 |
More Information
Set Membership: |
|

Genomic Map