Assay Details
Target Gene Details
Entrez Gene ID: | 20346 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A |
Gene Aliases: |
Hsema-I, SEMA1, SemD, Semad, coll-1 |
Location: |
Chr.5:13125371-13606234 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3a | NM_001243072.1 | NP_001230001.1 | ||
| NM_001243073.1 | NP_001230002.1 | |||
| NM_009152.4 | NP_033178.2 | |||
| AK140817.1 | BAE24487.1 | |||
| AK164242.1 | BAE37700.1 | |||
| BC057588.1 | AAH57588.1 | |||
| BC090844.1 | AAH90844.1 | |||
| D85028.2 | BAA19773.1 | |||
| L40484.1 | AAA73934.1 | |||
| L41541.1 | AAL77611.1 | |||
| X85993.1 | CAA59985.1 |
More Information
Set Membership: |
|

Genomic Map