Assay Details
Target Gene Details
Entrez Gene ID: | 20346 |
Gene Name: | sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A |
Gene Aliases: |
Hsema-I, SEMA1, SemD, Semad, coll-1 |
Location: |
Chr.5:13125371-13606234 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema3a | NM_001243072.1 | 9 | 1250 | NP_001230001.1 |
| NM_001243073.1 | 9 | 1090 | NP_001230002.1 | |
| NM_009152.4 | 8 | 1518 | NP_033178.2 | |
| AK140817.1 | 9 | 1091 | BAE24487.1 | |
| BC057588.1 | 8 | 1165 | AAH57588.1 | |
| BC090844.1 | 9 | 1250 | AAH90844.1 | |
| D85028.2 | 8 | 1520 | BAA19773.1 | |
| L40484.1 | 5 | 551 | AAA73934.1 | |
| L41541.1 | 8 | 869 | AAL77611.1 | |
| X85993.1 | 8 | 979 | CAA59985.1 |
More Information
Set Membership: |
|

Genomic Map