Assay Details
Target Gene Details
Entrez Gene ID: | 232413 |
Gene Name: | C-type lectin domain family 12, member a |
Gene Aliases: |
CLL-1, D230024O04, KLRL1, Micl |
Location: |
Chr.6:129349779-129365303 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec12a | NM_177686.4 | 2 | 316 | NP_808354.1 |
| AK084335.1 | 2 | 274 | BAC39163.1 | |
| BB850261.1 | 2 | 317 | ||
| BC094904.1 | 2 | 291 | AAH94904.1 |

Genomic Map