Assay Details
Target Gene Details
Entrez Gene ID: | 71183 |
Gene Name: | C-type lectin domain family 12, member B |
Gene Aliases: |
4933425B16Rik |
Location: |
Chr.6:129375513-129427634 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clec12b | NM_001204223.1 | NP_001191152.1 | ||
| NM_027709.2 | NP_081985.1 | |||
| AK016908.1 | BAB30491.1 | |||
| BC117712.2 |

Genomic Map