Assay Details
Target Gene Details
Entrez Gene ID: | 77574 |
Gene Name: | TRPM8 channel-associated factor 1 |
Gene Aliases: |
2810407D09Rik, 3321401G04Rik, A230020K05Rik, AI848529, Fam115a |
Location: |
Chr.6:42667999-42710110 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcaf1 | NM_029930.2 | NP_084206.1 | ||
| AK014379.1 | BAC25433.1 | |||
| AK053509.1 | BAC35410.1 | |||
| AK076242.1 | ||||
| AK083917.1 | BAC39061.1 | |||
| AK132148.1 | BAE20997.1 | |||
| AK148593.1 | ||||
| AK173013.1 | BAD32291.1 | |||
| BC031718.1 | AAH31718.1 | |||
| BC043304.1 | AAH43304.1 |

Genomic Map