Assay Details
Target Gene Details
Entrez Gene ID: | 12332 |
Gene Name: | capping protein (actin filament), gelsolin-like |
Gene Aliases: |
gCap39, mbh1 |
Location: |
Chr.6:72544391-72562983 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Capg | NM_007599.4 | NP_031625.2 | ||
| AK159577.1 | BAE35198.1 | |||
| AK159736.1 | BAE35330.1 | |||
| AK165153.1 | BAE38052.1 | |||
| BU696808.1 |
More Information
Set Membership: |
|

Genomic Map