Assay Details
Target Gene Details
Entrez Gene ID: | 319552 |
Gene Name: | RIKEN cDNA B230216G23 gene |
Gene Aliases: |
Npq, Spx |
Location: |
Chr.6:142410186-142419740 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B230216G23Rik | NM_001242345.1 | NP_001229274.1 | ||
| AK043509.1 | ||||
| AK045625.1 |

Genomic Map