Assay Details
Target Gene Details
Entrez Gene ID: | 213819 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
Cas1, Cast1 |
Location: |
Chr.6:4600563-4643381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casd1 | NM_145398.2 | NP_663373.2 | ||
| AB091829.1 | BAC77246.1 | |||
| AK193530.1 | ||||
| AK200014.1 | ||||
| BC018542.1 | AAH18542.1 | |||
| BC038009.1 | AAH38009.1 | |||
| BC058953.1 | AAH58953.1 | |||
| BC125377.1 | ||||
| BC125379.1 |
More Information
Set Membership: |
|

Genomic Map