Assay Details
Target Gene Details
Entrez Gene ID: | 66797 |
Gene Name: | contactin associated protein-like 2 |
Gene Aliases: |
5430425M22Rik, Caspr2, mKIAA0868 |
Location: |
Chr.6:45060061-47301388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntnap2 | NM_001004357.2 | NP_001004357.2 | ||
| NM_025771.3 | NP_080047.1 | |||
| AK013139.1 | BAB28674.1 | |||
| AK017341.1 | ||||
| AK129232.1 | BAC98042.1 | |||
| BC029826.1 | AAH29826.1 | |||
| BC064467.1 | AAH64467.1 | |||
| BC103799.1 | AAI03800.1 | |||
| BY297045.1 |

Genomic Map