Assay Details
Target Gene Details
Entrez Gene ID: | 330428 |
Gene Name: | transmembrane protein 52B |
Gene Aliases: |
D630042F21Rik |
Location: |
Chr.6:129512546-129519227 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem52b | NM_001081186.1 | NP_001074655.1 | ||
| AK085389.1 | ||||
| AK085571.1 | ||||
| BC120661.1 | ||||
| BC120663.1 |
More Information
Set Membership: |
|

Genomic Map