Assay Details
Target Gene Details
Entrez Gene ID: | 320405 |
Gene Name: | Ca2+-dependent activator protein for secretion 2 |
Gene Aliases: |
A230044C21Rik, Caps-2, Caps2, Cpd2 |
Location: |
Chr.6:23262773-23839421 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps2 | NM_001252105.1 | 5 | 1283 | NP_001239034.1 |
| NM_001252106.1 | 5 | 1283 | NP_001239035.1 | |
| NM_001252107.1 | 5 | 1283 | NP_001239036.1 | |
| NM_001252108.1 | 5 | 1283 | NP_001239037.1 | |
| NM_001252110.1 | 5 | 1283 | NP_001239039.1 | |
| NM_153163.4 | 5 | 1283 | NP_694803.3 | |
| AB098623.1 | 5 | 1282 | BAD05017.1 | |
| AB291947.1 | 5 | 1098 | ||
| AB291948.1 | 5 | 1168 | ||
| AB291949.1 | 5 | 1137 | ||
| AB291950.1 | 5 | 1168 | ||
| AF000969.2 | 5 | 909 | AAB58720.2 | |
| AK038568.1 | 5 | 1088 | BAC30047.1 | |
| AK046596.1 | 5 | 1129 | BAC32801.1 | |
| AK163203.1 | 5 | 1163 | ||
| AY072800.1 | 6 | 912 | AAL67937.1 | |
| BC047394.1 | 5 | 911 | AAH47394.1 | |
| BC055462.1 | 1 | 76 | AAH55462.1 | |
| BC059274.1 | 4 | 632 | AAH59274.1 | |
| BC080854.1 | 4 | 596 | AAH80854.1 |
More Information
Set Membership: |
|

Genomic Map