Assay Details
Target Gene Details
Entrez Gene ID: | 403088 |
Gene Name: | TRPM8 channel-associated factor 3 |
Gene Aliases: |
Eapa2, Fam115e |
Location: |
Chr.6:42587213-42597372 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcaf3 | NM_203396.1 | 5 | 2031 | NP_981933.1 |
| AY528666.1 | 5 | 2031 | AAS18676.1 |

Genomic Map