Assay Details
Target Gene Details
Entrez Gene ID: | 70020 |
Gene Name: | INO80 complex subunit B |
Gene Aliases: |
2510009I23Rik, HMG1YL4, Hmga1l4, Papa1, Znhit4 |
Location: |
Chr.6:83121828-83125029 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80b | NM_023547.1 | 2 | 157 | NP_076036.1 |
AK010946.1 | 2 | 259 | BAB27284.1 | |
BC019383.1 | 2 | 191 | AAH19383.1 | |
BC047990.1 | 2 | 192 | AAH47990.1 | |
BC058788.1 | 2 | 203 | AAH58788.1 |
