Assay Details
Target Gene Details
Entrez Gene ID: | 67529 |
Gene Name: | FGFR1 oncogene partner 2 |
Gene Aliases: |
1500031J01Rik |
Location: |
Chr.6:146577908-146599198 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr1op2 | NM_026218.2 | NP_080494.1 | ||
| AB041650.1 | 2 | 409 | ||
| AK004662.1 | BAB23452.1 | |||
| AK005320.1 | BAB23951.1 | |||
| AK008983.1 | BAB26006.1 | |||
| AK050102.1 | ||||
| AK151318.1 | BAE30299.1 | |||
| AK152617.1 | BAE31360.1 | |||
| AK153137.1 | BAE31749.1 | |||
| AK159805.1 | BAE35385.1 | |||
| AK169380.1 | BAE41127.1 | |||
| AK169383.1 | BAE41130.1 | |||
| BC028876.1 | AAH28876.1 |

Genomic Map