Assay Details
Target Gene Details
Entrez Gene ID: | 27055 |
Gene Name: | FK506 binding protein 9 |
Gene Aliases: |
1810014L23, AW549641, FKBP-63, FKBP-9, FKBP60, FKBP63, FKBP65RS |
Location: |
Chr.6:56832059-56879360 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp9 | NM_012056.2 | NP_036186.2 | ||
| AF090334.1 | AAC72964.1 | |||
| AF279263.1 | AAF79215.1 | |||
| AK007499.1 | BAB25071.1 | |||
| AK189046.1 | ||||
| BC026133.1 | AAH26133.1 | |||
| BC043129.1 | AAH43129.1 |

Genomic Map