Assay Details
Target Gene Details
Entrez Gene ID: | 58991 |
Gene Name: | ghrelin |
Gene Aliases: |
2210006E23Rik, Ghr, MTLRP, MTLRPAP, m46 |
Location: |
Chr.6:113716119-113727485 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ghrl | NM_021488.5 | NP_067463.2 | ||
| FJ355944.1 |
More Information
Set Membership: |
|

Genomic Map