Assay Details
Target Gene Details
Entrez Gene ID: | 58991 |
Gene Name: | ghrelin |
Gene Aliases: |
2210006E23Rik, Ghr, MTLRP, MTLRPAP, m46 |
Location: |
Chr.6:113716119-113727485 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ghrl | NM_001286404.1 | 2 | 186 | NP_001273333.1 |
| NM_001286405.1 | 2 | 189 | NP_001273334.1 | |
| NM_001286406.1 | NP_001273335.1 | |||
| NM_021488.5 | 3 | 237 | NP_067463.2 | |
| AB035701.1 | 2 | 179 | BAB19046.1 | |
| AB111891.1 | BAC77409.1 | |||
| AJ243503.1 | 2 | 202 | CAB46500.1 | |
| AK008658.1 | 3 | 207 | BAB25814.1 | |
| AK008860.1 | 3 | 207 | BAB25934.1 | |
| AY179430.1 | 2 | 179 | AAO27350.1 | |
| BC132230.1 | 2 | 203 | ||
| BC132232.1 | 2 | 203 | ||
| DQ993169.1 | ||||
| FJ914266.1 | 2 | 243 |
More Information
Set Membership: |
|

Genomic Map