Assay Details
Target Gene Details
Entrez Gene ID: | 75379 |
Gene Name: | RIKEN cDNA 4930599N23 gene |
Gene Aliases: |
- |
Location: |
Chr.6:39118473-39148312 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930599N23Rik | NR_045813.1 | |||
| AK016417.1 | BAC25481.1 |
Target Gene Details
Entrez Gene ID: | 338523 |
Gene Name: | lysine (K)-specific demethylase 7A |
Gene Aliases: |
A630082K20Rik, BB041802, ENSMUSG00000073143, Jhdm1d, mKIAA1718 |
Location: |
Chr.6:39136620-39206773 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm7a | NM_001033430.4 | NP_001028602.2 | ||
| AK035529.1 | ||||
| AK129429.1 | BAC98239.1 | |||
| AK136169.1 | ||||
| AK136209.1 | BAE22876.1 | |||
| AK136238.1 | BAE22890.1 | |||
| BC007161.1 | AAH07161.1 | |||
| BC145848.1 |
More Information
Set Membership: |
|

Genomic Map