Assay Details
Target Gene Details
Entrez Gene ID: | 108078 |
Gene Name: | oxidized low density lipoprotein (lectin-like) receptor 1 |
Gene Aliases: |
LOX-1, SR-EI, Scare1 |
Location: |
Chr.6:129485247-129507165 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olr1 | NM_001301094.1 | NP_001288023.1 | ||
| NM_001301096.1 | NP_001288025.1 | |||
| NM_138648.2 | NP_619589.2 | |||
| AF303744.1 | AAG44998.1 | |||
| AK154687.1 | BAE32764.1 | |||
| AK160919.1 | ||||
| HQ603085.1 | ||||
| HQ603086.1 |

Genomic Map