Assay Details
Target Gene Details
Entrez Gene ID: | 12304 |
Gene Name: | protein disulfide isomerase associated 4 |
Gene Aliases: |
AI987846, Cai, ERp-72, Erp72 |
Location: |
Chr.6:47796141-47813512 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia4 | NM_009787.2 | 5 | 917 | NP_033917.2 |
| AK028292.1 | 5 | 767 | BAC25863.1 | |
| AK146288.1 | 5 | 750 | BAE27045.1 | |
| AK148801.1 | 5 | 837 | ||
| AK150566.1 | 5 | 798 | BAE29664.1 | |
| AK161534.1 | 5 | 755 | BAE36446.1 | |
| AK168447.1 | 5 | 721 | ||
| AK169387.1 | 5 | 762 | BAE41134.1 | |
| BC066857.1 | 4 | 516 | AAH66857.1 | |
| BC141078.1 | 5 | 813 | ||
| J05186.1 | 5 | 800 | AAA39907.1 | |
| Y00884.1 | 4 | 623 | CAA68777.1 |
More Information
Set Membership: |
|

Genomic Map