Assay Details
Target Gene Details
Entrez Gene ID: | 12304 |
Gene Name: | protein disulfide isomerase associated 4 |
Gene Aliases: |
AI987846, Cai, ERp-72, Erp72 |
Location: |
Chr.6:47796141-47813512 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdia4 | NM_009787.2 | NP_033917.2 | ||
| AK028292.1 | BAC25863.1 | |||
| AK146288.1 | BAE27045.1 | |||
| AK148801.1 | ||||
| AK150566.1 | BAE29664.1 | |||
| AK161534.1 | BAE36446.1 | |||
| AK168447.1 | ||||
| AK169387.1 | BAE41134.1 | |||
| BC066857.1 | AAH66857.1 | |||
| BC141078.1 | ||||
| J05186.1 | AAA39907.1 | |||
| Y00884.1 | CAA68777.1 |
More Information
Set Membership: |
|

Genomic Map