Assay Details
Target Gene Details
Entrez Gene ID: | 68202 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5 |
Gene Aliases: |
2900002J19Rik |
Location: |
Chr.6:24518662-24527687 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
More Information
Set Membership: |
|

Genomic Map