Assay Details
Target Gene Details
Entrez Gene ID: | 68198 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2 |
Gene Aliases: |
1810011O01Rik, 8kDa, AGGG, AI325567, CI-AGGG |
Location: |
Chr.6:39592551-39599472 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb2 | NM_026612.3 | NP_080888.1 | ||
| AK007495.1 | BAB25068.1 | |||
| AK010642.1 | BAB27084.1 | |||
| AK019008.1 | BAB31508.1 | |||
| AK216480.1 | ||||
| BC013510.1 | AAH13510.1 |
More Information
Set Membership: |
|

Genomic Map