Assay Details
Target Gene Details
Entrez Gene ID: | 14823 |
Gene Name: | glutamate receptor, metabotropic 8 |
Gene Aliases: |
A230002O04, Gprc1h, mGluR8 |
Location: |
Chr.6:27275121-28135837 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm8 | NM_008174.2 | NP_032200.2 | ||
| AK020963.1 | ||||
| AK038395.1 | ||||
| AK165623.1 | ||||
| AY673682.1 | AAT76980.1 | |||
| BC051384.1 | ||||
| BC137596.1 | ||||
| BC144753.1 | ||||
| BC144754.1 | ||||
| U17252.1 | AAA68149.1 |
More Information
Set Membership: |
|

Genomic Map