Assay Details
Target Gene Details
Entrez Gene ID: | 114142 |
Gene Name: | forkhead box P2 |
Gene Aliases: |
2810043D05Rik, AI449000, CAG-16, D0Kist7 |
Location: |
Chr.6:14901226-15441977 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxp2 | NM_001286607.1 | 11 | 1576 | NP_001273536.1 |
| NM_053242.4 | 13 | 1976 | NP_444472.2 | |
| NM_212435.1 | 11 | 1639 | NP_997600.1 | |
| AF339106.1 | 10 | 1317 | AAK69651.1 | |
| AK051668.1 | 13 | 1959 | BAC34711.1 | |
| AK082361.1 | 13 | 1648 | BAC38477.1 | |
| AY079003.1 | 13 | 1605 | AAL85482.1 | |
| BC058960.1 | 11 | 1573 | AAH58960.1 | |
| BC062926.1 | 11 | 1639 | AAH62926.1 |

Genomic Map