Assay Details
Target Gene Details
Entrez Gene ID: | 19051 |
Gene Name: | protein phosphatase 1, regulatory subunit 17 |
Gene Aliases: |
Gsbs, Ppp1r2 |
Location: |
Chr.6:56017515-56032688 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r17 | NM_011153.3 | NP_035283.1 | ||
| AF071562.1 | AAD12589.1 | |||
| AF500906.1 | AAM21706.1 | |||
| AK078335.1 | BAC37224.1 | |||
| BC026822.1 | AAH26822.1 |
More Information
Set Membership: |
|

Genomic Map