Assay Details
Target Gene Details
Entrez Gene ID: | 16438 |
Gene Name: | inositol 1,4,5-trisphosphate receptor 1 |
Gene Aliases: |
D6Pas2, ENSMUSG00000072853, Gm10429, IP3R1, InsP3R, Ip3r, Itpr-1, P400, Pcp-1, Pcp1, opt |
Location: |
Chr.6:108213083-108551116 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 50 - Exon 51 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpr1 | NM_010585.5 | NP_034715.3 | ||
| BC003271.1 | AAH03271.1 | |||
| BY145967.1 | ||||
| X15373.1 | CAA33433.1 |
Target Gene Details
Entrez Gene ID: | 102465766 |
Gene Name: | microRNA 7661 |
Gene Aliases: |
mmu-mir-7661 |
Location: |
Chr.6:108489743-108489803 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7661 | NR_106121.1 |
More Information
Set Membership: |
|

Genomic Map