Assay Details
Target Gene Details
Entrez Gene ID: | 56284 |
Gene Name: | mitochondrial ribosomal protein L19 |
Gene Aliases: |
9030416F12Rik, 9130412E02Rik, D6Ertd157e, MRP-L15, RLX1, Rpml15 |
Location: |
Chr.6:81957826-81965949 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl19 | NM_026490.2 | 1 | 167 | NP_080766.1 |
| AK021180.1 | 1 | 161 | ||
| AK030793.1 | 1 | 155 | ||
| AK050637.1 | 1 | 166 | BAC34357.1 | |
| AK141554.1 | 1 | 177 | BAE24733.1 | |
| BC020315.2 | 1 | 143 | AAH20315.1 | |
| BC043921.1 | 1 | 148 | AAH43921.1 | |
| BC061064.1 | 1 | 151 | AAH61064.1 |
More Information
Set Membership: |
|

Genomic Map