Assay Details
Target Gene Details
Entrez Gene ID: | 56284 |
Gene Name: | mitochondrial ribosomal protein L19 |
Gene Aliases: |
9030416F12Rik, 9130412E02Rik, D6Ertd157e, MRP-L15, RLX1, Rpml15 |
Location: |
Chr.6:81957826-81965949 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl19 | NM_026490.2 | NP_080766.1 | ||
| AK021180.1 | ||||
| AK030793.1 | ||||
| AK050637.1 | BAC34357.1 | |||
| AK141554.1 | BAE24733.1 | |||
| BC020315.2 | AAH20315.1 | |||
| BC043921.1 | AAH43921.1 | |||
| BC061064.1 | AAH61064.1 |
More Information
Set Membership: |
|

Genomic Map