Assay Details
Target Gene Details
Entrez Gene ID: | 108017 |
Gene Name: | FXYD domain-containing ion transport regulator 4 |
Gene Aliases: |
0610008I02Rik, AI267073, Chif |
Location: |
Chr.6:117933559-117938510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fxyd4 | NM_001173372.1 | NP_001166843.1 | ||
| NM_033648.1 | NP_387468.1 | |||
| AK018728.1 | BAB31372.1 | |||
| AU041511.1 | ||||
| BC086918.1 | AAH86918.1 |
More Information
Set Membership: |
|

Genomic Map