Assay Details
Target Gene Details
Entrez Gene ID: | 66356 |
Gene Name: | lysine rich nucleolar protein 1 |
Gene Aliases: |
2310008H09Rik, Tsg118 |
Location: |
Chr.7:118842217-118855998 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Knop1 | NM_001168218.1 | NP_001161690.1 | ||
| NM_001168219.1 | NP_001161691.1 | |||
| NM_001168220.1 | NP_001161692.1 | |||
| NM_023197.3 | NP_075686.2 | |||
| AF034580.1 | AAD01984.1 | |||
| AK009230.1 | BAB26153.1 | |||
| AK083845.1 | BAC39038.1 | |||
| BC055941.1 | AAH55941.1 | |||
| CD553078.1 | ||||
| CV557281.1 |
More Information
Set Membership: |
|

Genomic Map