Assay Details
Target Gene Details
Entrez Gene ID: | 54486 |
Gene Name: | hematopoietic prostaglandin D synthase |
Gene Aliases: |
H-PGDS, Ptgds2 |
Location: |
Chr.6:65117293-65145024 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hpgds | NM_019455.4 | NP_062328.3 | ||
| AK020246.1 | BAB32037.1 | |||
| AK039392.1 | BAC30336.1 | |||
| BC116735.1 | ||||
| BC116737.1 | ||||
| D82072.1 | BAA96845.1 |
More Information
Set Membership: |
|

Genomic Map