Assay Details
Target Gene Details
Entrez Gene ID: | 21848 |
Gene Name: | tripartite motif-containing 24 |
Gene Aliases: |
A130082H20Rik, AI447469, D430004I05Rik, TIF1, TIF1-alpha, TIF1alpha, Tif1a |
Location: |
Chr.6:37869880-37968445 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trim24 | NM_001272064.1 | NP_001258993.1 | ||
| NM_001272076.1 | NP_001259005.1 | |||
| NM_145076.4 | NP_659542.3 | |||
| AB221637.1 | ||||
| AK080890.1 | ||||
| AK084870.1 | ||||
| AK133144.1 | BAE21530.1 | |||
| BC056959.1 | AAH56959.1 | |||
| S78219.1 | AAB34289.1 | |||
| S78221.1 | AAB34290.1 |
More Information
Set Membership: |
|

Genomic Map