Assay Details
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zyx | NM_001289617.1 | NP_001276546.1 | ||
| NM_001289618.1 | NP_001276547.1 | |||
| NM_001289619.1 | NP_001276548.1 | |||
| NM_011777.3 | NP_035907.1 | |||
| AK031415.1 | BAC27394.1 | |||
| AK035743.1 | BAC29173.1 | |||
| AK076491.1 | ||||
| AK090110.1 | ||||
| AK147812.1 | BAE28154.1 | |||
| AK161709.1 | ||||
| AK165290.1 | 5 | 1432 | ||
| AK170571.1 | BAE41886.1 | |||
| AK217332.1 | ||||
| BC054775.1 | AAH54775.1 | |||
| X99063.1 | CAA67510.1 | |||
| Y07711.1 | CAA68984.1 |
More Information
Set Membership: |
|

Genomic Map