Assay Details
Target Gene Details
Entrez Gene ID: | 69683 |
Gene Name: | ER membrane protein complex subunit 10 |
Gene Aliases: |
2310044H10Rik, 5430410O10Rik, Inm02, Mirta22 |
Location: |
Chr.7:44489938-44496524 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc10 | NM_197991.2 | NP_932108.2 | ||
| AK009801.1 | ||||
| AK150499.1 | ||||
| AK160274.1 | ||||
| AK169010.1 | BAE40807.1 | |||
| AK171657.1 | BAE42592.1 | |||
| AK183758.1 | ||||
| AK189635.1 | ||||
| AY761096.1 | AAV30544.1 | |||
| AY761098.1 | AAV30546.1 | |||
| BC006065.1 | AAH06065.1 | |||
| BC020179.1 | AAH20179.1 |
More Information
Set Membership: |
|

Genomic Map