Assay Details
Target Gene Details
Entrez Gene ID: | 24110 |
Gene Name: | ubiquitin specific peptidase 18 |
Gene Aliases: |
1110058H21Rik, AW047653, UBP43, Ubp15 |
Location: |
Chr.6:121245906-121270917 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp18 | NM_011909.2 | NP_036039.2 | ||
| AF069502.1 | AAD21222.1 | |||
| AK004296.1 | ||||
| AK149772.1 | BAE29076.1 | |||
| AK151011.1 | BAE30032.1 | |||
| AK151260.1 | BAE30248.1 | |||
| AK151722.1 | BAE30640.1 | |||
| AK152034.1 | BAE30893.1 | |||
| AK152574.1 | BAE31326.1 | |||
| AK153312.1 | BAE31893.1 | |||
| BC138562.1 | ||||
| BC138577.1 | ||||
| CT010228.1 | CAJ18436.1 |
More Information
Set Membership: |
|

Genomic Map