Assay Details
Target Gene Details
| Entrez Gene ID: | 66508 | 
| Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 1 | 
| Gene Aliases: | 2400001E08Rik, Pdro, p18 | 
| Location: | Chr.7:101905837-101911903 on Build GRCm38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Lamtor1 | NM_025605.3 | 1 | 60 | NP_079881.2 | 
| AK010247.1 | ||||
| AK013389.1 | BAB28825.1 | |||
| AK144476.1 | BAE25909.1 | |||
| BC020142.1 | AAH20142.1 | |||
| BC126973.1 | 
Target Gene Details
| Entrez Gene ID: | 791260 | 
| Gene Name: | transmembrane O-methyltransferase | 
| Gene Aliases: | Comt2, F930017I19Rik | 
| Location: | Chr.7:101899803-101906359 on Build GRCm38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Tomt | NM_001081679.1 | NP_001075148.1 | ||
| DQ854743.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top