Assay Details
Target Gene Details
Entrez Gene ID: | 436022 |
Gene Name: | dynein, axonemal assembly factor 3 |
Gene Aliases: |
6030429G01Rik, b2b1739Clo |
Location: |
Chr.7:4522948-4532488 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnaaf3 | NM_001033548.2 | NP_001028720.1 | ||
| AK134335.1 | BAE22103.1 | |||
| BC150983.1 |
More Information
Set Membership: |
|

Genomic Map