Assay Details
Target Gene Details
Entrez Gene ID: | 321008 |
Gene Name: | RIKEN cDNA 6330408A02 gene |
Gene Aliases: |
- |
Location: |
Chr.7:13258967-13278721 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 6330408A02Rik | NM_177312.4 | NP_796286.2 | ||
| AK018141.1 | BAB31091.1 | |||
| AK028865.1 | ||||
| AK162673.1 | BAE37016.1 | |||
| CV783961.1 |
Target Gene Details
Entrez Gene ID: | 16881 |
Gene Name: | ligase I, DNA, ATP-dependent |
Gene Aliases: |
AL033288, LigI |
Location: |
Chr.7:13277313-13311427 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lig1 | NM_001199310.1 | NP_001186239.1 | ||
| BC138541.1 | ||||
| BC138542.1 | ||||
| U19604.1 | AAB60500.1 |
More Information
Set Membership: |
|

Genomic Map