Assay Details
Target Gene Details
Entrez Gene ID: | 67148 |
Gene Name: | family with sequence similarity 103, member A1 |
Gene Aliases: |
2410047I02Rik, 2610204K14Rik, RAM |
Location: |
Chr.7:81762953-81769490 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam103a1 | NM_025997.2 | NP_080273.1 | ||
| AK010694.1 | BAB27126.1 | |||
| AK011881.1 | BAB27895.1 | |||
| AK018614.1 | BAB31308.1 | |||
| AK079819.1 | BAC37755.1 | |||
| AK081301.1 | BAC38188.1 | |||
| AK085230.1 | BAC39395.1 | |||
| AK150938.1 | BAE29973.1 | |||
| AK160347.1 | BAE35751.1 | |||
| AK169392.1 | BAE41139.1 | |||
| BC058367.1 | AAH58367.1 | |||
| BC091760.1 | AAH91760.1 | |||
| BC096399.1 | AAH96399.1 |

Genomic Map