Assay Details
Target Gene Details
Entrez Gene ID: | 269831 |
Gene Name: | tetraspanin 12 |
Gene Aliases: |
9030619E17, AI426782, AI663988, AW111457, Tm4sf12 |
Location: |
Chr.6:21771391-21853349 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan12 | NM_173007.3 | 3 | 332 | NP_766595.1 |
| AK033554.1 | 2 | 167 | BAC28354.1 | |
| AK050737.1 | 3 | 333 | BAC34399.1 | |
| BC065152.1 | 2 | 143 | AAH65152.1 | |
| BC068240.1 | 2 | 194 | AAH68240.1 |

Genomic Map