Assay Details
Target Gene Details
Entrez Gene ID: | 66870 |
Gene Name: | serpine1 mRNA binding protein 1 |
Gene Aliases: |
1200009K13Rik, 9330147J08Rik, AL022786, Pairbp1 |
Location: |
Chr.6:67266979-67289302 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serbp1 | NM_001113564.1 | 5 | 862 | NP_001107036.1 |
| NM_001113565.1 | NP_001107037.1 | |||
| NM_001113566.1 | NP_001107038.1 | |||
| NM_025814.2 | 5 | 880 | NP_080090.2 | |
| AK004678.1 | 5 | 880 | BAB23466.1 | |
| AK010851.1 | 5 | 850 | ||
| AK010860.1 | BAB27229.1 | |||
| AK011051.1 | 5 | 849 | ||
| AK045190.1 | 4 | 1434 | BAC32255.1 | |
| AK088278.1 | BAC40253.1 | |||
| AK131990.1 | BAE20924.1 | |||
| AK135473.1 | ||||
| AK139670.1 | ||||
| AK144764.1 | 5 | 860 | BAE26054.1 | |
| AK146418.1 | BAE27153.1 | |||
| AK149505.1 | BAE28925.1 | |||
| BC013665.1 | AAH13665.1 | |||
| BC030502.1 | AAH30502.1 |
More Information
Set Membership: |
|

Genomic Map