Assay Details
Target Gene Details
Entrez Gene ID: | 66870 |
Gene Name: | serpine1 mRNA binding protein 1 |
Gene Aliases: |
1200009K13Rik, 9330147J08Rik, AL022786, Pairbp1 |
Location: |
Chr.6:67266979-67289302 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serbp1 | NM_001113564.1 | 6 | 964 | NP_001107036.1 |
| NM_001113565.1 | 6 | 937 | NP_001107037.1 | |
| NM_001113566.1 | 6 | 919 | NP_001107038.1 | |
| NM_025814.2 | 6 | 982 | NP_080090.2 | |
| AK004678.1 | 6 | 982 | BAB23466.1 | |
| AK010851.1 | 6 | 952 | ||
| AK010860.1 | 6 | 937 | BAB27229.1 | |
| AK011051.1 | 6 | 951 | ||
| AK088278.1 | 6 | 888 | BAC40253.1 | |
| AK131990.1 | 6 | 888 | BAE20924.1 | |
| AK135473.1 | 5 | 719 | ||
| AK144764.1 | 6 | 962 | BAE26054.1 | |
| AK146418.1 | 6 | 906 | BAE27153.1 | |
| AK149505.1 | 6 | 891 | BAE28925.1 | |
| BC013665.1 | 6 | 873 | AAH13665.1 | |
| BC030502.1 | 6 | 881 | AAH30502.1 |
More Information
Set Membership: |
|

Genomic Map