Assay Details
Target Gene Details
Entrez Gene ID: | 264134 |
Gene Name: | tetratricopeptide repeat domain 26 |
Gene Aliases: |
9330141E21Rik, 9430097H08Rik |
Location: |
Chr.6:38381474-38427647 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc26 | NM_153600.2 | NP_705828.2 | ||
| AK035185.1 | BAC28974.1 | |||
| BC031513.1 | AAH31513.1 | |||
| BC066060.1 | AAH66060.1 |

Genomic Map