Assay Details
Target Gene Details
Entrez Gene ID: | 100861870 |
Gene Name: | predicted gene, 21284 |
Gene Aliases: |
- |
Location: |
Chr.6:83568039-83588126 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21284 | NR_078345.1 | |||
| AK137133.1 |
Target Gene Details
Entrez Gene ID: | 70527 |
Gene Name: | STAM binding protein |
Gene Aliases: |
5330424L14Rik, 5730422L11Rik, AW107289, Amsh, mKIAA4198 |
Location: |
Chr.6:83543206-83575770 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stambp | NM_024239.2 | NP_077201.1 | ||
| AB010123.1 | BAB78604.1 | |||
| AK017600.1 | BAB30832.1 | |||
| AK019907.1 | BAB31909.1 | |||
| AK046995.1 | ||||
| AK053000.1 | 1 | 56 | ||
| AK136961.1 | BAE23188.1 | |||
| AK139391.1 | BAE23991.1 | |||
| AK168810.1 | BAE40637.1 | |||
| AK220541.1 | BAD90533.1 | |||
| BC003497.1 | AAH03497.1 | |||
| BC006939.1 | AAH06939.1 | |||
| BC025111.1 | AAH25111.1 | |||
| CJ074803.1 |
More Information
Set Membership: |
|

Genomic Map