Assay Details
Target Gene Details
Entrez Gene ID: | 67290 |
Gene Name: | RIKEN cDNA 3110040N11 gene |
Gene Aliases: |
AI851475 |
Location: |
Chr.7:81780283-81790462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110040N11Rik | NM_026077.3 | NP_080353.1 | ||
| AK014163.1 | BAB29184.1 | |||
| AK018003.1 | BAB31031.1 | |||
| AK019261.1 | BAB31634.1 | |||
| AK170168.1 | BAE41614.1 | |||
| BC027500.1 | AAH27500.1 |

Genomic Map