Assay Details
Target Gene Details
Entrez Gene ID: | 21991 |
Gene Name: | triosephosphate isomerase 1 |
Gene Aliases: |
AI255506, Tpi, Tpi-1 |
Location: |
Chr.6:124810592-124814296 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpi1 | NM_009415.2 | NP_033441.2 | ||
| AK002270.1 | ||||
| AK010808.1 | BAB27194.1 | |||
| AK146013.1 | BAE26832.1 | |||
| AK149768.1 | BAE29073.1 | |||
| AK150735.1 | BAE29810.1 | |||
| AK159741.1 | BAE35334.1 | |||
| AK167437.1 | BAE39523.1 | |||
| AK168060.1 | ||||
| AK168446.1 | BAE40350.1 | |||
| AK168756.1 | BAE40594.1 | |||
| BC046761.1 | AAH46761.1 | |||
| BU945752.1 | ||||
| X53333.1 | CAA37420.1 |
More Information
Set Membership: |
|

Genomic Map